Assay Details
| Gene Symbol: itprid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079133.2 | NP_001072601.2 | 12 - 13 | 3105 | 65 | |
| XM_018097149.1 | XP_017952638.1 | 12 - 13 | 2798 | 65 | ||
| GenBank mRNA | BC122975.1 | - | 12 - 13 | 3052 | 65 | |
| DT419235.1 | - | 2 - 3 | 469 | 65 | ||
More Information
Set Membership: |
|

itprid2
Genomic Map