Assay Details
| Gene Symbol: itprid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079133.2 | NP_001072601.2 | 13 - 14 | 3231 | 70 | |
| XM_018097149.1 | XP_017952638.1 | 13 - 14 | 2924 | 70 | ||
| GenBank mRNA | BC122975.1 | - | 13 - 14 | 3178 | 70 | |
| DT419235.1 | - | 3 - 4 | 595 | 70 | ||
| DT424483.1 | - | 2 - 3 | 152 | 70 | ||
More Information
Set Membership: |
|

itprid2
Genomic Map