Assay Details
| Gene Symbol: itprid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079133.2 | NP_001072601.2 | 16 - 17 | 3877 | 89 | |
| XM_018097149.1 | XP_017952638.1 | 16 - 17 | 3570 | 89 | ||
| GenBank mRNA | BC122975.1 | - | 16 - 17 | 3824 | 89 | |
| CX449743.2 | - | 2 - 3 | 569 | 89 | ||
| DT424483.1 | - | 5 - 6 | 798 | 89 | ||
More Information
Set Membership: |
|

itprid2
Genomic Map