Assay Details
| Gene Symbol: lox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079143.1 | NP_001072611.1 | 1 - 2 | 766 | 72 | |
| XM_012965635.2 | XP_012821089.1 | 1 - 2 | 760 | 72 | ||
| XM_012965639.1 | XP_012821093.1 | 1 - 2 | 760 | 72 | ||
| GenBank mRNA | BC122993.1 | - | 1 - 2 | 766 | 72 | |
More Information
Set Membership: |
|

lox
Genomic Map