Assay Details
Gene Symbol: larp6-like.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079168.1 | NP_001072636.1 | 3 - 4 | 411 | 99 | |
GenBank mRNA | BC123089.1 | - | 3 - 4 | 411 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
