Assay Details
| Gene Symbol: prcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079171.1 | NP_001072639.1 | 1 - 2 | 256 | 67 | |
| GenBank mRNA | BC123912.1 | - | 1 - 2 | 256 | 67 | |
More Information
Set Membership: |
|

prcp
Genomic Map