Assay Details
| Gene Symbol: naip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079203.1 | NP_001072671.1 | 5 - 6 | 1273 | 95 | |
| XM_002934832.4 | XP_002934878.3 | 29 - 30 | 8229 | 95 | ||
| GenBank mRNA | BC123992.1 | - | 5 - 6 | 1273 | 95 | |
More Information
Set Membership: |
|

naip
Genomic Map