Assay Details
Gene Symbol: thbs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079205.1 | NP_001072673.1 | 20 - 21 | 2809 | 56 | |
XM_012965779.2 | XP_012821233.1 | 21 - 22 | 3081 | 56 | ||
GenBank mRNA | BC124000.1 | - | 20 - 21 | 2809 | 56 | |
BC135149.1 | - | 20 - 21 | 2740 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
