Assay Details
| Gene Symbol: pdxk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079245.1 | NP_001072713.1 | 922 | 56 | ||
| GenBank mRNA | BC125667.1 | - | 922 | 56 | ||
| CT030525.1 | - | 5 - 6 | 802 | 56 | ||
More Information
Set Membership: |
|

pdxk
Genomic Map