Assay Details
Gene Symbol: mpz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079273.2 | NP_001072741.1 | 1 - 2 | 215 | 71 | |
NM_001358944.1 | NP_001345873.1 | 1 - 2 | 215 | 71 | ||
NM_001358946.1 | NP_001345875.1 | 1 - 2 | 215 | 71 | ||
GenBank mRNA | BC125729.1 | - | 1 - 2 | 215 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
