Assay Details
Gene Symbol: zfp36l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079290.1 | NP_001072758.1 | 1 - 2 | 282 | 85 | |
GenBank mRNA | BC125786.1 | - | 1 - 2 | 282 | 85 | |
CT025463.2 | - | 1 - 2 | 337 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
