Assay Details
| Gene Symbol: rgs5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079294.1 | NP_001072762.1 | 3 - 4 | 330 | 68 | |
| GenBank mRNA | BC125794.1 | - | 3 - 4 | 330 | 68 | |
More Information
Set Membership: |
|

rgs5
Genomic Map