Assay Details
Gene Symbol: hsp90aa1.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079297.1 | NP_001072765.1 | 10 - 11 | 2097 | 104 | |
GenBank mRNA | BC125798.1 | - | 10 - 11 | 2097 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
