Assay Details
| Gene Symbol: cib2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079342.1 | NP_001072810.1 | 3 - 4 | 263 | 97 | |
| XM_012966762.1 | XP_012822216.1 | 3 - 4 | 223 | 97 | ||
| GenBank mRNA | BC121478.1 | - | 3 - 4 | 263 | 97 | |
More Information
Set Membership: |
|

cib2
Genomic Map