Assay Details
| Gene Symbol: ifngr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079346.2 | NP_001072814.2 | 6 - 7 | 1020 | 68 | |
| GenBank mRNA | BC121592.1 | - | 6 - 7 | 996 | 68 | |
| BC167921.1 | - | 6 - 7 | 961 | 68 | ||
| CR848177.2 | - | 6 - 7 | 990 | 68 | ||
More Information
Set Membership: |
|

ifngr1
Genomic Map