Assay Details
Gene Symbol: nr1h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079385.1 | NP_001072853.1 | 5 - 6 | 698 | 60 | |
XM_012966763.2 | XP_012822217.1 | 5 - 6 | 945 | 60 | ||
GenBank mRNA | BC122937.1 | - | 5 - 6 | 698 | 60 | |
CT025497.2 | - | 5 - 6 | 691 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
