Assay Details
| Gene Symbol: eef1a1o | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203970.3 | NP_989301.1 | 6 - 7 | 1393 | 79 | |
| GenBank mRNA | BC064177.1 | - | 6 - 7 | 1340 | 79 | |
| BC157320.1 | - | 6 - 7 | 1301 | 79 | ||
| BC158425.1 | - | 6 - 7 | 1297 | 79 | ||
| CR761806.2 | - | 6 - 7 | 1267 | 79 | ||
More Information
Set Membership: |
|

eef1a1o
Genomic Map