Assay Details
| Gene Symbol: ppm1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016158.3 | NP_001016158.2 | 3 - 4 | 1284 | 66 | |
| XM_012968369.2 | XP_012823823.1 | 3 - 4 | 1153 | 66 | ||
| GenBank mRNA | BC135714.1 | - | 3 - 4 | 1284 | 66 | |
More Information
Set Membership: |
|

ppm1a
Genomic Map