Assay Details
Gene Symbol: nrbf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016183.3 | NP_001016183.1 | 3 - 4 | 287 | 80 | |
GenBank mRNA | BC118696.1 | - | 3 - 4 | 287 | 80 | |
CR761828.2 | - | 3 - 4 | 314 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
