Assay Details
| Gene Symbol: fezf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016305.3 | NP_001016305.1 | 2 - 3 | 1040 | 79 | |
| XM_012961082.2 | XP_012816536.1 | 1 - 2 | 1061 | 79 | ||
| GenBank mRNA | BC136188.1 | - | 2 - 3 | 1040 | 79 | |
| CR761501.2 | - | 1 - 2 | 793 | 79 | ||
More Information
Set Membership: |
|

fezf2
Genomic Map