Assay Details
Gene Symbol: erg28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016539.3 | NP_001016539.1 | 212 | 82 | ||
GenBank mRNA | BC135223.1 | - | 212 | 82 | ||
CR760767.2 | - | 1 - 2 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
