Assay Details
Gene Symbol: erg28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016539.3 | NP_001016539.1 | 301 | 70 | ||
GenBank mRNA | BC135223.1 | - | 301 | 70 | ||
CR760767.2 | - | 2 - 3 | 301 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
