Assay Details
Gene Symbol: msantd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016739.3 | NP_001016739.1 | 2 - 3 | 572 | 80 | |
XM_012964722.2 | XP_012820176.1 | 2 - 3 | 607 | 80 | ||
GenBank mRNA | BC135254.1 | - | 2 - 3 | 572 | 80 | |
CR760001.2 | - | 2 - 3 | 568 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
