Assay Details
Gene Symbol: msantd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016739.3 | NP_001016739.1 | 4 - 5 | 846 | 105 | |
XM_012964722.2 | XP_012820176.1 | 4 - 5 | 881 | 105 | ||
XM_018094659.1 | XP_017950148.1 | 3 - 4 | 739 | 105 | ||
GenBank mRNA | BC135254.1 | - | 4 - 5 | 846 | 105 | |
CR760001.2 | - | 4 - 5 | 842 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
