Assay Details
Gene Symbol: plpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016858.4 | NP_001016858.3 | 4 - 5 | 964 | 71 | |
XM_012961143.2 | XP_012816597.1 | 4 - 5 | 1597 | 71 | ||
GenBank mRNA | AL635807.2 | - | 1 - 2 | 358 | 71 | |
BC135252.1 | - | 4 - 5 | 964 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
