Assay Details
| Gene Symbol: cox5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017049.3 | NP_001017049.1 | 3 - 4 | 438 | 95 | |
| GenBank mRNA | BC135615.1 | - | 3 - 4 | 438 | 95 | |
| CR761816.2 | - | 3 - 4 | 351 | 95 | ||
More Information
Set Membership: |
|

cox5a
Genomic Map