Assay Details
Gene Symbol: lzic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017291.3 | NP_001017291.1 | 3 - 4 | 269 | 77 | |
XM_012966659.2 | XP_012822113.1 | 3 - 4 | 377 | 77 | ||
XM_012966660.2 | XP_012822114.1 | 3 - 4 | 816 | 77 | ||
XM_012966661.2 | XP_012822115.1 | 3 - 4 | 387 | 77 | ||
XM_018095400.1 | XP_017950889.1 | 3 - 4 | 377 | 77 | ||
GenBank mRNA | BC135363.1 | - | 3 - 4 | 269 | 77 | |
CR855656.2 | - | 3 - 4 | 275 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
