Assay Details
Gene Symbol: nsa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017326.3 | NP_001017326.1 | 4 - 5 | 658 | 73 | |
GenBank mRNA | BC135791.1 | - | 4 - 5 | 658 | 73 | |
CR848589.2 | - | 3 - 4 | 529 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
