Assay Details
| Gene Symbol: tmem205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044477.2 | NP_001037942.1 | 3 - 4 | 337 | 58 | |
| XM_012953169.2 | XP_012808623.1 | 4 - 5 | 508 | 58 | ||
| XM_012953170.2 | XP_012808624.1 | 3 - 4 | 465 | 58 | ||
| XM_012953171.2 | XP_012808625.1 | 3 - 4 | 394 | 58 | ||
| GenBank mRNA | BC136112.1 | - | 3 - 4 | 337 | 58 | |
| CR761405.2 | - | 3 - 4 | 303 | 58 | ||
More Information
Set Membership: |
|

tmem205
Genomic Map