Assay Details
| Gene Symbol: timm13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045772.1 | NP_001039237.1 | 1 - 2 | 127 | 84 | |
| GenBank mRNA | BC160996.1 | - | 1 - 2 | 170 | 84 | |
| CR942737.2 | - | 1 - 2 | 127 | 84 | ||
More Information
Set Membership: |
|

timm13
Genomic Map