Assay Details
Gene Symbol: guca1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079039.1 | NP_001072507.1 | 1 - 2 | 682 | 74 | |
GenBank mRNA | BC121947.1 | - | 1 - 2 | 682 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
