Assay Details
Gene Symbol: chst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079061.1 | NP_001072529.1 | 2 - 3 | 718 | 99 | |
XM_018093095.1 | XP_017948584.1 | 3 - 4 | 652 | 99 | ||
XM_018093096.1 | XP_017948585.1 | 1 - 2 | 48 | 218 | ||
GenBank mRNA | BC121984.1 | - | 2 - 3 | 718 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018093096.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
