Assay Details
| Gene Symbol: srd5a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079071.1 | NP_001072539.1 | 4 - 5 | 851 | 96 | |
| XM_012965419.2 | XP_012820873.1 | 5 - 6 | 978 | 96 | ||
| XM_012965424.2 | XP_012820878.1 | 5 - 6 | 947 | 96 | ||
| XM_012965426.2 | XP_012820880.1 | 4 - 5 | 788 | 96 | ||
| GenBank mRNA | BC122039.1 | - | 4 - 5 | 851 | 96 | |
| CU025196.1 | - | 5 - 6 | 948 | 96 | ||
More Information
Set Membership: |
|

srd5a3
Genomic Map