Assay Details
| Gene Symbol: cenpu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079077.1 | NP_001072545.1 | 195 | 63 | ||
| GenBank mRNA | BC122049.1 | - | 195 | 63 | ||
| CR848437.2 | - | 161 | 63 | |||
More Information
Set Membership: |
|

cenpu
Genomic Map