Assay Details
Gene Symbol: dhrs12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079079.1 | NP_001072547.1 | 1 - 2 | 58 | 75 | |
GenBank mRNA | BC122052.1 | - | 1 - 2 | 58 | 75 | |
CT030358.1 | - | 1 - 2 | 157 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
