Assay Details
Gene Symbol: tmem246 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079081.1 | NP_001072549.1 | 1 | 1356 | 82 | |
XM_012965452.2 | XP_012820906.1 | 2 | 1544 | 82 | ||
XM_012965459.2 | XP_012820913.1 | 2 | 1510 | 82 | ||
XM_012965461.1 | XP_012820915.1 | 2 | 1854 | 82 | ||
XM_012965466.2 | XP_012820920.1 | 2 | 1387 | 82 | ||
XM_018095439.1 | XP_017950928.1 | 2 | 1548 | 82 | ||
GenBank mRNA | BC122061.1 | - | 1 | 1356 | 82 | |
CU075399.1 | - | 1 | 1251 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
