Assay Details
Gene Symbol: b3glct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079083.1 | NP_001072551.1 | 7 - 8 | 638 | 63 | |
XM_012957278.2 | XP_012812732.1 | 6 - 7 | 731 | 63 | ||
XM_018091349.1 | XP_017946838.1 | 5 - 6 | 462 | 63 | ||
GenBank mRNA | BC122064.1 | - | 7 - 8 | 638 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
