Assay Details
| Gene Symbol: spon2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079092.2 | NP_001072560.2 | 665 | 66 | ||
| GenBank mRNA | DR870839.1 | - | 2 - 3 | 296 | 66 | |
| DR872131.1 | - | 4 - 5 | 665 | 66 | ||
More Information
Set Membership: |
|

spon2
Genomic Map