Assay Details
Gene Symbol: tmem164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079093.1 | NP_001072561.1 | 1 - 2 | 816 | 56 | |
GenBank mRNA | BC122081.1 | - | 1 - 2 | 816 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
