Assay Details
| Gene Symbol: duoxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079095.1 | NP_001072563.1 | 4 - 5 | 392 | 98 | |
| GenBank mRNA | BC122088.1 | - | 4 - 5 | 392 | 98 | |
More Information
Set Membership: |
|

duoxa1
Genomic Map