Assay Details
| Gene Symbol: tmcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079127.1 | NP_001072595.1 | 1 - 2 | 658 | 61 | |
| XM_012965579.2 | XP_012821033.1 | 2 - 3 | 499 | 61 | ||
| XM_012965582.2 | XP_012821036.1 | 2 - 3 | 422 | 61 | ||
| XM_012965585.1 | XP_012821039.1 | 2 - 3 | 226 | 61 | ||
| GenBank mRNA | BC122963.1 | - | 1 - 2 | 658 | 61 | |
More Information
Set Membership: |
|

tmcc2
Genomic Map