Assay Details
Gene Symbol: galnt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079237.2 | NP_001072705.1 | 3 - 4 | 555 | 69 | |
GenBank mRNA | BC124051.1 | - | 2 - 3 | 472 | 69 | |
BC135911.1 | - | 3 - 4 | 555 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
