Assay Details
| Gene Symbol: crip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079267.1 | NP_001072735.1 | 1 - 2 | 87 | 58 | |
| XM_012968693.2 | XP_012824147.1 | 1 - 2 | 308 | 58 | ||
| XM_012968694.2 | XP_012824148.1 | 1 - 2 | 305 | 58 | ||
| GenBank mRNA | BC125715.1 | - | 1 - 2 | 87 | 58 | |
More Information
Set Membership: |
|

crip2
Genomic Map