Assay Details
| Gene Symbol: ccdc92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079326.1 | NP_001072794.1 | 3 - 4 | 496 | 83 | |
| XM_012965970.2 | XP_012821424.1 | 3 - 4 | 935 | 83 | ||
| XM_012965977.2 | XP_012821431.1 | 3 - 4 | 548 | 83 | ||
| XM_012965986.2 | XP_012821440.1 | 3 - 4 | 554 | 83 | ||
| XM_012965996.2 | XP_012821450.1 | 3 - 4 | 551 | 83 | ||
| GenBank mRNA | BC121331.1 | - | 3 - 4 | 496 | 83 | |
| CU075491.1 | - | 3 - 4 | 416 | 83 | ||
More Information
Set Membership: |
|

ccdc92
Genomic Map