Assay Details
| Gene Symbol: rragd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079428.1 | NP_001072896.1 | 4 - 5 | 816 | 73 | |
| GenBank mRNA | BC118790.1 | - | 4 - 5 | 816 | 73 | |
More Information
Set Membership: |
|

rragd
Genomic Map