Assay Details
| Gene Symbol: sox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080996.1 | NP_001074465.1 | 1 | 1452 | 56 | |
| GenBank mRNA | BC121846.1 | - | 1 | 405 | 56 | |
| BC159121.1 | - | 1 | 1326 | 56 | ||
| EF192052.1 | - | 1 | 1452 | 56 | ||
More Information
Set Membership: |
|

sox1
Genomic Map