Assay Details
| Gene Symbol: rfx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083343.1 | NP_001076812.1 | 677 | 70 | ||
| GenBank mRNA | BC064876.1 | - | 648 | 70 | ||
| BC135604.1 | - | 677 | 70 | |||
More Information
Set Membership: |
|

rfx5
Genomic Map