Assay Details
| Gene Symbol: fitm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097185.1 | NP_001090654.1 | 1 - 2 | 379 | 71 | |
| XM_012971476.2 | XP_012826930.1 | 2 - 3 | 535 | 71 | ||
| XM_012971477.2 | XP_012826931.1 | 2 - 3 | 517 | 71 | ||
| XM_012971478.2 | XP_012826932.1 | 2 - 3 | 521 | 71 | ||
| XM_012971479.2 | XP_012826933.1 | 2 - 3 | 486 | 71 | ||
| XM_012971480.2 | XP_012826934.1 | 2 - 3 | 442 | 71 | ||
| XM_012971481.2 | XP_012826935.1 | 2 - 3 | 541 | 71 | ||
| GenBank mRNA | BC127286.1 | - | 1 - 2 | 379 | 71 | |
More Information
Set Membership: |
|

fitm2
Genomic Map