Assay Details
Gene Symbol: pak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097202.1 | NP_001090671.1 | 5 - 6 | 1532 | 72 | |
XM_012967790.2 | XP_012823244.1 | 5 - 6 | 2397 | 72 | ||
XM_012967791.2 | XP_012823245.1 | 5 - 6 | 2035 | 72 | ||
XM_018096079.1 | XP_017951568.1 | 5 - 6 | 2119 | 72 | ||
GenBank mRNA | BC127316.1 | - | 5 - 6 | 1532 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
