Assay Details
Gene Symbol: dusp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097242.1 | NP_001090711.1 | 6 - 7 | 1214 | 74 | |
XM_012958346.2 | XP_012813800.1 | 6 - 7 | 1274 | 74 | ||
GenBank mRNA | BC128637.1 | - | 6 - 7 | 1214 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
